Learn More
5-Ethylpyridine-2-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Ethyl-pyridine-2-carboxylic Acid |
| CAS | 770-08-1 |
| Molecular Formula | C8 H9 N O2 |
| Quantity | 500 mg |
| Synonym | 2-Pyridinecarboxylic acid, 5-ethyl-, Picolinic acid, 5-ethyl- (6CI,7CI,8CI), 5-Ethyl-2-pyridinecarboxylic acid, 5-Ethyl-2-picolinic acid, 5-Ethylpicolinic acid, 5-Ethylpyridine-2-carboxylic acid |
| InChI Formula | InChI=1S/C8H9NO2/c1-2-6-3-4-7(8(10)11)9-5-6/h3-5H,2H2,1H3,(H,10,11) |
| SMILES | CCc1ccc(nc1)C(=O)O |
| IUPAC Name | 5-ethylpyridine-2-carboxylic acid |
| Molecular Weight (g/mol) | 151.16 |
| Formula Weight | 151.0633 |