Learn More
5-Dimethylamino-thiophene-2-carbaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Dimethylamino-thiophene-2-carbaldehyde |
| CAS | 24372-46-1 |
| Molecular Formula | C7H9NOS |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C7H9NOS/c1-8(2)7-4-3-6(5-9)10-7/h3-5H,1-2H3 |
| SMILES | CN(C)c1ccc(C=O)s1 |
| IUPAC Name | 5-(dimethylamino)thiophene-2-carbaldehyde |
| Molecular Weight (g/mol) | 155.22 |
| Formula Weight | 155.0405 |