Learn More
5-Delta-Bromobutylhydantoin, TRC
Specifications
Specifications
| Chemical Name or Material | 5-delta-Bromobutylhydantoin |
| CAS | 28484-49-3 |
| Molecular Formula | C7 H11 Br N2 O2 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C7H11BrN2O2/c8-4-2-1-3-5-6(11)10-7(12)9-5/h5H,1-4H2,(H2,9,10,11,12) |
| SMILES | BrCCCCC1NC(=O)NC1=O |
| IUPAC Name | 5-(4-bromobutyl)imidazolidine-2,4-dione |
| Molecular Weight (g/mol) | 235.08 |
| Formula Weight | 234.0004 |