missing translation for 'onlineSavingsMsg'
Learn More

5-(chlorosulfonyl)-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 10656630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10656630 5 g Glass Bottle 5g
10714682 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656630 Supplier Thermo Scientific Acros Supplier No. 438010050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37098-75-2
InChI Key CISMZCVQNVNWJW-UHFFFAOYSA-N
Synonym 5-chlorosulfonyl-2-fluorobenzoic acid, 2-fluoro-5-chlorosulfonyl benzoic acid, 5-chlorosulphonyl-2-fluorobenzoic acid, 5-chlorosulfonyl-2-fluoro-benzoic acid, benzoic acid, 5-chlorosulfonyl-2-fluoro, 5-chlorosulfonyl-2-fluorobenzoicacid, acmc-1afaw, ksc577o0r, 2-fluoro-5-chlorosulfonylbenzoic acid
PubChem CID 608984
IUPAC Name 5-chlorosulfonyl-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(=O)O)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 111.0°C to 113.0°C
Color White
Elemental Analysis 33.82 to 36.64% C, 1.62 to 1.76% H
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C7H4ClFO4S
Quantity 5 g
Molecular Weight (g/mol) 238.613
Formula Weight 238.62
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-(chlorosulfonyl)-2-fluorobenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.