missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-m-phenylenediamine, 97%

Product Code. 11342536
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11342536

Brand: Thermo Scientific Alfa Aesar L06485.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33786-89-9 | C6H7ClN2 | 142.586 g/mol

5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33786-89-9
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00014775
InChI Key VZNUCJOYPXKLTA-UHFFFAOYSA-N
Synonym 5-chloro-m-phenylenediamine, 3,5-diaminochlorobenzene, 5-chloro-1,3-benzenediamine, 1,3-benzenediamine, 5-chloro, 5-chloro-1,3-phenylenediamine, ccris 2886, 5-amino-3-chloroaniline, acmc-20am42, 2-13-00-00029 beilstein handbook reference, 5-chloro-1,3-diaminobenzene
PubChem CID 73335
IUPAC Name 5-chlorobenzene-1,3-diamine
SMILES C1=C(C=C(C=C1N)Cl)N

Specifications

Melting Point 99°C to 103°C
Quantity 25 g
UN Number UN1673
Beilstein 3029854
Solubility Information Slightly soluble in water.
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-m-phenylenediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.