missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-furaldehyde, 98%, stab. with 2% ethanol

Product Code. 15489109
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15489109 Supplier Thermo Scientific Alfa Aesar Supplier No. L19971.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 21508-19-0 | C5H3ClO2 | 130.527 g/mol

(+)-1-Deoxynojirimycin hydrochloride interferes with normal processing of N-linked glycoproteins, producing reduced amounts of complex type oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-1-Deoxynojirimycin hydrochloride interferes with normal processing of N-linked glycoproteins, producing reduced amounts of complex type oligosaccharides.

Solubility
Insoluble in water.

Notes
Air
TRUSTED_SUSTAINABILITY

Specifications

CAS 21508-19-0
Melting Point 33°C to 36°C
Boiling Point 70°C (10 mmHg)
Molecular Formula C5H3ClO2
MDL Number MFCD02752599
Quantity 1 g
Beilstein 108401
Sensitivity Air sensitive
Synonym 5-chloro-2-furaldehyde, 5-chlorofurfural, 2-furancarboxaldehyde, 5-chloro, 5-chloro-furfural, 2-chlorofuran-5-aldehyde, acmc-1cq7z, 5-chloro-furan-2carbaldehyde, 5-chlorofuran-2 carbaldehyde, 5-chloro-furan-2-carbaldehyde, 5-chlorofuran-2-carboxaldehyde
Solubility Information Insoluble in water.
InChI Key DGAUAVDWXYXXGQ-UHFFFAOYSA-N
SMILES C1=C(OC(=C1)Cl)C=O
IUPAC Name 5-chlorofuran-2-carbaldehyde
Molecular Weight (g/mol) 130.527
PubChem CID 2769630
Formula Weight 130.53
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-furaldehyde, stabilized with 2% ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.