Learn More
5-Chloro-1,3,4-thiadiazol-2-amine, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-1,3,4-thiadiazol-2-amine |
| CAS | 37566-40-8 |
| Molecular Formula | C2H2ClN3S |
| Quantity | 1 g |
| Synonym | 2-Amino-5-chloro-1,3,4-thiadiazole, NSC 231514 |
| InChI Formula | InChI=1S/C2H2ClN3S/c3-1-5-6-2(4)7-1/h(H2,4,6) |
| SMILES | Nc1nnc(Cl)s1 |
| IUPAC Name | 5-chloro-1,3,4-thiadiazol-2-amine |
| Molecular Weight (g/mol) | 135.58 |