Learn More
5-Carboxy SU 5402, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Carboxy-2-[(1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-4-methyl-1H-pyrrole-3-propanoic Acid |
| CAS | 258831-77-5 |
| Molecular Formula | C18H16N2O5 |
| Quantity | 10 mg |
| Synonym | 5-Carboxy SU 5402, 5-Carboxy-2-[(1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-4-methyl-1H-pyrrole-3-propanoic Acid |
| InChI Formula | InChI=1S/C18H16N2O5/c1-9-10(6-7-15(21)22)14(19-16(9)18(24)25)8-12-11-4-2-3-5-13(11)20-17(12)23/h2-5,8,19H,6-7H2,1H3,(H,20,23)(H,21,22)(H,24,25)/b12-8- |
| SMILES | Cc1c(CCC(=O)O)c(\C=C\2/C(=O)Nc3ccccc23)[nH]c1C(=O)O |
| IUPAC Name | 4-(2-carboxyethyl)-3-methyl-5-[(Z)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrole-2-carboxylic acid |
| Molecular Weight (g/mol) | 340.33 |
| Formula Weight | 340.1059 |