missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyridine-2-carboxylic acid, 98%

Product Code. 11461697
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11461697

Brand: Thermo Scientific Alfa Aesar B25675.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30766-11-1 | C6H3BrNO2 | 201.00 g/mol

5-Bromopyridine-2-carboxylic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromopyridine-2-carboxylic acid is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30766-11-1
Molecular Formula C6H3BrNO2
Molecular Weight (g/mol) 201.00
MDL Number MFCD00234149
InChI Key MNNQIBXLAHVDDL-UHFFFAOYSA-M
Synonym 5-bromopicolinic acid, 5-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo, 5-bromo-2-carboxypyridine, 5-bromo-pyridine-2-carboxylic acid, 5-bromo-2-picolinic acid, 5-bromo-2-carboxy pyridine, 5-bromo-2-pyridine carboxylic acid, 5-bromopyridine-2-carboxylicacid
PubChem CID 608544
IUPAC Name 5-bromopyridine-2-carboxylic acid
SMILES [O-]C(=O)C1=CC=C(Br)C=N1

Specifications

Melting Point 173°C to 175°C
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 202.01
Percent Purity 98%
Chemical Name or Material 5-Bromopyridine-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.