Learn More
5-Bromo-4-fluoro-1,3-benzodioxole, 96%, Thermo Scientific™
Product Code. 15489388
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15489388
Brand: Thermo Scientific Alfa Aesar H62946.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1226808-75-8
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD16295053
InChI Key FMEWVPPYMHXEGT-UHFFFAOYSA-N
Synonym 5-bromo-4-fluorobenzo d 1,3 dioxole, 5-bromo-4-fluoro-2h-1,3-benzodioxole, acmc-209am4, 5-bromo-4-fluoro-1,3-benzodioxole
PubChem CID 53216233
IUPAC Name 5-bromo-4-fluoro-1,3-benzodioxole
SMILES C1OC2=C(O1)C(=C(C=C2)Br)F
Specifications
Quantity 250 mg
Formula Weight 219.01
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Bromo-4-fluoro-1,3-benzodioxole