Learn More
5-Bromo-4-chloro-3-indolyl-1,3-diacetate, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Bromo-4-chloro-3-indolyl-1,3-diacetate |
| CAS | 3030-06-6 |
| Molecular Formula | C12 H9 Br Cl N O3 |
| Quantity | 1 g |
| Synonym | Ethanone, 1-[3-(acetyloxy)-5-bromo-4-chloro-1H-indol-1-yl]-, 1H-Indol-3-ol, 1-acetyl-5-bromo-4-chloro-, acetate (ester) (9CI), Indol-3-ol, 1-acetyl-5-bromo-4-chloro-, acetate (7CI), Indol-3-ol, 1-acetyl-5-bromo-4-chloro-, acetate (ester) (8CI), Indoxyl, 1-acetyl-5-bromo-4-chloro-, acetate (6CI), 1-[3-(Acetyloxy)-5-bromo-4-chloro-1H-indol-1-yl]ethanone, 1-Acetyl-5-bromo-4-chloro-1H-indol-3-yl acetate, 5-Bromo-4-chloro-3-indolyl-1,3-diacetate |
| InChI Formula | InChI=1S/C12H9BrClNO3/c1-6(16)15-5-10(18-7(2)17)11-9(15)4-3-8(13)12(11)14/h3-5H,1-2H3 |
| SMILES | CC(=O)Oc1cn(C(=O)C)c2ccc(Br)c(Cl)c12 |
| IUPAC Name | (1-acetyl-5-bromo-4-chloroindol-3-yl) acetate |
| Molecular Weight (g/mol) | 330.56 |
| Formula Weight | 328.9454 |