Learn More
5-Bromo-2-phenoxypyrimidine, 97%, BLD Pharm™
Product Code. 30948214
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948214 1 g 1g
30947160 25 g 25g
30948336 250 mg 250mg
30945997 100 g 100g
30942356 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30948214 Supplier BLD Pharm Supplier No. BD1028281g
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 257280-25-4
Molecular Formula C10H7BrN2O
MDL Number MFCD03646346
Quantity 1 g
SMILES BrC1=CN=C(N=C1)OC2=CC=CC=C2
Molecular Weight (g/mol) 251.08
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-phenoxypyrimidine