Learn More
5-Bromo-2-hydroxypyrimidine, TRC
Product Code. 30347933
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347933 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30347933 Supplier Toronto Research Chemicals Supplier No. TRCB6844751G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Bromo-2-hydroxypyrimidine
CAS 38353-06-9
Molecular Formula C4 H3 Br N2 O
Quantity 1 g
Synonym Macitentan Impurity 32
InChI Formula InChI=1S/C4H3BrN2O/c5-3-1-6-4(8)7-2-3/h1-2H,(H,6,7,8)
SMILES Oc1ncc(Br)cn1
IUPAC Name 5-bromopyrimidin-2-ol
Molecular Weight (g/mol) 174.98
Formula Weight 173.9429