Learn More
5-Bromo-2,3-dihydro-1,4-benzodioxane, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 58328-39-5 |
| Molecular Formula | C8H7BrO2 |
| MDL Number | MFCD03095029 |
| Quantity | 10 g |
| SMILES | BrC1=C2OCCOC2=CC=C1 |
| Molecular Weight (g/mol) | 215.04 |
| Percent Purity | 96% |
| Chemical Name or Material | 5-Bromo-2,3-dihydro-1,4-benzodioxane |