missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino Salicylic Acid Methyl Ester-13C6, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | 5-Amino Salicylic Acid Methyl Ester-13C6 |
| Molecular Formula | 13C6 C2 H9 N O3 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C8H9NO3/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4,10H,9H2,1H3/i2+1,3+1,4+1,5+1,6+1,7+1 |
| SMILES | COC(=O)[13c]1[13cH][13c](N)[13cH][13cH][13c]1O |
| IUPAC Name | methyl 5-amino-2-hydroxy(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-triene-1-carboxylate |
| Molecular Weight (g/mol) | 173.118 |
| Formula Weight | 173.078 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction