missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-methylisoxazole, 98%

Product Code. 11409673
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11409673

Brand: Thermo Scientific Alfa Aesar A12446.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14678-02-5 | C4H6N2O | 98.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14678-02-5
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00003151
InChI Key FNXYWHTZDAVRTB-UHFFFAOYSA-N
Synonym 5-amino-3-methylisoxazole, 3-methylisoxazol-5-amine, 5-isoxazolamine, 3-methyl, 3-methyl-5-isoxazolamine, unii-0g66r1h4rw, 3-methyl-isoxazol-5-ylamine, 3-methylisoxazole-5-ylamine, 3-methylisoxazole-5-amine, acmc-209yk8, 3-methylisooxazole-5-amine
PubChem CID 84590
IUPAC Name 3-methyl-1,2-oxazol-5-amine
SMILES CC1=NOC(=C1)N

Specifications

Melting Point 81°C to 85°C
Quantity 25 g
Beilstein 107906
Formula Weight 98.11
Percent Purity ≥98%
Chemical Name or Material 5-Amino-3-methylisoxazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.