Learn More
5-Amino-2-(trifluoromethyl)benzimidazole, 96%, Thermo Scientific™
Product Code. 11986731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11986731 5 g 5g
11976731 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11986731 Supplier Thermo Scientific Acros Supplier No. 442340050
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Chemical Identifiers
CAS 3671-66-7
Molecular Formula C8H6F3N3
Molecular Weight (g/mol) 201.15
InChI Key CKEKFQLHCAZGSP-UHFFFAOYSA-N
Synonym 5-amino-2-trifluoromethyl benzimidazole, 1h-benzimidazol-5-amine, 2-trifluoromethyl, 2-trifluoromethyl-1h-benzo d imidazol-5-amine, benzimidazole, 5-amino-2-trifluoromethyl, 2-trifluoromethyl-1h-benzimidazol-5-amine, 2-trifluoromethyl-1h-1,3-benzodiazol-5-amine, 5-amino-2-trifluoromethylbenzimidazole, 2-trifluoromethyl benzimidazole-5-ylamine, 2-trifluoromethyl-1h-benzimidazol-6-amine, 2-trifluoromethyl-3h-benzoimidazol-5-ylamine
PubChem CID 19345
IUPAC Name 2-(trifluoromethyl)-3H-benzimidazol-5-amine
SMILES C1=CC2=C(C=C1N)NC(=N2)C(F)(F)F
Specifications
Melting Point 189.0°C to 191.0°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 201.15
Percent Purity 96%
Chemical Name or Material 5-Amino-2-(trifluoromethyl)benzimidazole
compliance-icons
Product Identifier
  • 5-Amino-2-(trifluoromethyl)benzimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.