Learn More
5-Amino-2-naphthalenol, TRC
Product Code. 30317418
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317418 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30317418 Supplier Toronto Research Chemicals Supplier No. TRCA6186605G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Amino-2-naphthalenol
CAS 86-97-5
Molecular Formula C10 H9 N O
Quantity 5 g
Synonym 5-Amino-2-naphthol, 1-Amino-6-hydroxynaphthaline, 1-Amino-6-naphthol, 5-Amino-2-hydroxynaphthalene, 5-Amino-2-naphthol, 6-Hydroxy-1-naphthylamine, NSC 4012
InChI Formula InChI=1S/C10H9NO/c11-10-3-1-2-7-6-8(12)4-5-9(7)10/h1-6,12H,11H2
SMILES Nc1cccc2cc(O)ccc12
IUPAC Name 5-aminonaphthalen-2-ol
Molecular Weight (g/mol) 159.185
Formula Weight 159.068