Learn More
5-Amino-2-chloro-2,3-dihydrothiazolo[4,5-d]pyrimidine-7-(6H)-one, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Amino-2-chloro-2,3-dihydrothiazolo[4,5-d]pyrimidine-7-(6H) |
| CAS | 30161-95-6 |
| Molecular Formula | C5 H5 Cl N4 O S |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C5H5ClN4OS/c6-4-8-2-1(12-4)3(11)10-5(7)9-2/h4H,(H4,7,8,9,10,11) |
| SMILES | NC1=NC2=C(SC(Cl)N2)C(=O)N1 |
| IUPAC Name | 5-amino-2-chloro-3,6-dihydro-2H-[1,3]thiazolo[4,5-d]pyrimidin-7-one |
| Molecular Weight (g/mol) | 204.64 |
| Formula Weight | 203.9873 |