Learn More
5-Amino-1-naphthol, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Amino-1-naphthol |
| CAS | 83-55-6 |
| Molecular Formula | C10H9NO |
| Quantity | 100 mg |
| Synonym | 1-Amino-5-hydroxynaphthalene, 1-Amino-5-naphthol, 5-Amino-1-naphthalenol, 5-Amino-1-naphthol, 5-Amino-α-naphthol, 5-Hydroxy-1-naphthalenamine, 5-Hydroxy-1-naphthylamine, C.I. 76650, NSC 1499 |
| InChI Formula | InChI=1S/C10H9NO/c11-9-5-1-4-8-7(9)3-2-6-10(8)12/h1-6,12H,11H2 |
| SMILES | OC1=C2C=CC=C(N)C2=CC=C1 |
| IUPAC Name | 5-aminonaphthalen-1-ol |
| Molecular Weight (g/mol) | 159.18 |
| Formula Weight | 159.0684 |