Learn More
5-Acetylthiophene-2-carbonitrile, 97%, Thermo Scientific™
Product Code. 10556423
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
5g
Product Code. 10556423
Brand: Thermo Scientific Maybridge TL00354EA
Request Bulk or Custom Format
Chemical Identifiers
CAS 88653-55-8
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
InChI Key VSHPLUBHIUFLES-UHFFFAOYSA-N
Synonym 2-acetyl-5-cyanothiophene, 2-acetyl-5-cyanotiophene, 5-cyano-2-acetylthiophene, 5-acetyl-2-thiophenecarbonitrile, maybridge1_000101, acmc-20ao65, 2-cyano-5-acetyl-thiophene, 5-acetyl-thiophene-2-carbonitrile, 5-ethanoylthiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-acetyl-7ci,9ci
PubChem CID 2747565
IUPAC Name 5-acetylthiophene-2-carbonitrile
SMILES CC(=O)C1=CC=C(S1)C#N
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 151.19
Percent Purity 97%
Chemical Name or Material 5-Acetylthiophene-2-carbonitrile