missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dihydrouracil-5,5,6,6-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Dihydro Uracil-d4 |
| CAS | 334473-41-5 |
| Molecular Formula | C4 D4 H2 N2 O2 |
| Quantity | 0.01 g |
| Synonym | 2,4(1 H,3 H)-Pyrimidinedione-5,6-d2, dihydro-d2- (9 CI), Dihydro-d2-2,4(1 H,3 H)-pyrimidinedione-5,6-d2, Hydrouracil-d4, 5,6-Dihydro-2,4(1 H,3 H)-pyrimidinedione-5,5,6,6-d4, 5,6-Dihydro-2,4-dihydroxypyrimidine-5,5,6,6-d4, 5,6-Dihydrouracil-d4, 5,6-Dihydrouracil-5,5,6,6-D4, Dihydrouracil-d4 |
| InChI Formula | InChI=1 S/C4H6N2O2/c7-3-1-2-5-4(8)6-3/h1-2H2,(H2,5,6,7,8)/i1D2,2D2 |
| SMILES | [2 H]C1([2 H])NC(=O)NC(=O)C1([2 H])[2 H] |
| IUPAC Name | 5,5,6,6-tetradeuterio-1,3-diazinane-2,4-dione |
| Molecular Weight (g/mol) | 118.127 |
| Formula Weight | 118.068 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction