missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro-4h-Pyrrolo[3,4-D]Thiazole Dihydrochloride, TRC

Product Code. 30372908
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372908 100 mg -
30377364 10 mg 10mg
30371223 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372908 Supplier Toronto Research Chemicals Supplier No. TRCD679593100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393686-96-8
Molecular Formula C5H6N2S.2HCl
Molecular Weight (g/mol) 199.1
Synonym 4H-Pyrrolo[3,4-d]thiazole, 5,6-dihydro-, hydrochloride (1:2) (ACI), 5,6-Dihydro-4H-pyrrolo[3,4-d]thiazole dihydrochloride
SMILES Cl.Cl.C1NCc2scnc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H6N2S.2ClH/c1-4-5(2-6-1)8-3-7-4;;/h3,6H,1-2H2;2*1H
Formula Weight 197.9785
Chemical Name or Material 5,6-Dihydro-4h-Pyrrolo[3,4-D]Thiazole Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.