Learn More
5,6,7,8-Tetrahydro-1-naphthylamine, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 5,6,7,8-Tetrahydro-1-naphthylamine |
| CAS | 2217-41-6 |
| Molecular Formula | C10H13N |
| Quantity | 2.5 g |
| Synonym | 1,2,3,4-Tetrahydro-5-aminonaphthalene, 1,2,3,4-Tetrahydro-5-naphthalenamine, 1,2,3,4-Tetrahydro-5-naphthylamine, 1-Amino-5,6,7,8-tetrahydronaphthalene, 5,6,7,8-Tetrahydro-1-naphthalenamine, 5,6,7,8-Tetrahydro-1-naphthalenylamine, 5-Amino-1,2,3,4-tetrahydronaphthalene, 5-Aminotetralin, 5-Tetralinylamine, NSC 53503 |
| InChI Formula | InChI=1S/C10H13N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h3,5,7H,1-2,4,6,11H2 |
| SMILES | Nc1cccc2CCCCc12 |
| IUPAC Name | 5,6,7,8-tetrahydronaphthalen-1-amine |
| Molecular Weight (g/mol) | 147.22 |
| Formula Weight | 147.1048 |