Learn More
5,5-Dibromobarbituric acid, 95+%
Product Code. 11966541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11966541 1 g 1g
11976541 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
Product Code. 11966541 Supplier Thermo Scientific Acros Supplier No. 439840010
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
CAS: 511-67-1 | C4H2Br2N2O3 | 285.88 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 511-67-1
Molecular Formula C4H2Br2N2O3
Molecular Weight (g/mol) 285.88
MDL Number MFCD00010595
InChI Key AMATXUCYHHHHHB-UHFFFAOYSA-N
Synonym 5,5-dibromobarbituric acid, dibromin, 5,5-dibromopyrimidine-2,4,6 1h,3h,5h-trione, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dibromo, dibrombarbitursaure, dibromobarbituric acid, acmc-20am7j, barbituric acid,5-dibromo, diethyl trimethylsilylphosphate
PubChem CID 94877
IUPAC Name 5,5-dibromo-1,3-diazinane-2,4,6-trione
SMILES BrC1(Br)C(=O)NC(=O)NC1=O
Specifications
Melting Point 240.0°C to 242.0°C
Quantity 1 g
Formula Weight 285.88
Chemical Name or Material 5,5-Dibromobarbituric acid
compliance-icons
Product Identifier
  • 5,5-Dibromobarbituric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.