Learn More
(5,5’-Bis(2-(4-(benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6,6’-dichloro-3-hydroxy-3,3’-biindoline-2,2’-dione), TRC
Specifications
Specifications
| Chemical Name or Material | 5,5'-Bis(2-(4-(benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6,6'-dichloro-3-hydroxy-3,3'-biindoline-2,2'-dione |
| CAS | 1303996-68-0 |
| Molecular Formula | C42H40Cl2N8O3S2 |
| Quantity | 25 mg |
| Synonym | [3,3'-Bi-2H-indole]-2,2'-dione, 5,5'-bis[2-[4-(1,2-benzisothiazol-3-yl)-1-piperazinyl]ethyl]-6,6'-dichloro-1,1',3,3'-tetrahydro-3-hydroxy-, 5,5'-Bis[2[4-(1,2-benzisothiazol-3-yl)piperazin-1-yl]ethyl]-6,6'-dichloro-3-hydroxy-1,1',3,3'-tetrahydro-2H,2H'-3,3'-biindole-2,2'-dione, Ziprasidone Related Compound C (USP) |
| InChI Formula | InChI=1S/C42H40Cl2N8O3S2/c43-31-23-33-29(21-25(31)9-11-49-13-17-51(18-14-49)38-27-5-1-3-7-35(27)56-47-38)37(40(53)45-33)42(55)30-22-26(32(44)24-34(30)46-41(42)54)10-12-50-15-19-52(20-16-50)39-28-6-2-4-8-36(28)57-48-39/h1-8,21-24,37,55H,9-20H2,(H,45,53)(H,46,54) |
| SMILES | OC1(C2C(=O)Nc3cc(Cl)c(CCN4CCN(CC4)c5nsc6ccccc56)cc23)C(=O)Nc7cc(Cl)c(CCN8CCN(CC8)c9nsc%10ccccc9%10)cc17 |
| IUPAC Name | 5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-3-[5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-2-oxo-1,3-dihydroindol-3-yl]-6-chloro-3-hydroxy-1H-indol-2-one |
| Molecular Weight (g/mol) | 839.9 |
| Formula Weight | 839.2 |