missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Methoxyphenoxy)pentanoic Acid, TRC

Product Code. 30473405
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473405 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473405 Supplier Toronto Research Chemicals Supplier No. TRCM26576025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(4-Methoxyphenoxy)pentanoic Acid
CAS 374565-57-8
Molecular Formula C12H16O4
Quantity 25 mg
InChI Formula InChI=1S/C12H16O4/c1-15-10-5-7-11(8-6-10)16-9-3-2-4-12(13)14/h5-8H,2-4,9H2,1H3,(H,13,14)
SMILES COc1ccc(OCCCCC(=O)O)cc1
IUPAC Name 5-(4-methoxyphenoxy)pentanoic acid
Molecular Weight (g/mol) 224.25
Formula Weight 224.1049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.