Learn More
5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide, 95%, BLD Pharm™
Product Code. 31432887
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31432887 5 g
31421693 100 mg
31431048 1 g
31427165 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Product Code. 31432887 missing translation for 'mfr' BLD Pharm missing translation for 'supplierNo' BD1167305g
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
CAS 66640-86-6
Molecular Formula C10H18N4O2S
MDL Number MFCD00078532
Quantity 5 g
SMILES NNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
Molecular Weight (g/mol) 258.34
Percent Purity 95%
Chemical Name or Material 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide