Learn More
5-(3-Methoxyphenyl)isoxazole, 95%, Thermo Scientific™
Product Code. 15496857
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. Quantity unitSize
15496857 10 g 10g
15486857 1 g 1g
2 options
Product Code. 15496857
Brand: Thermo Scientific Alfa Aesar H33965.09
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1194374-23-6
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD13176591
InChI Key FDZRTKRNZFFBEC-UHFFFAOYSA-N
Synonym 5-3-methoxyphenyl isoxazole, 5-3-methoxyphenyl-1,2-oxazole
PubChem CID 64984921
IUPAC Name 5-(3-methoxyphenyl)-1,2-oxazole
SMILES COC1=CC=CC(=C1)C2=CC=NO2
Specifications
Boiling Point 115°C to 117°C (0.46 mmHg)
Refractive Index 1.586
Quantity 10 g
Formula Weight 175.19
Percent Purity 95%
Chemical Name or Material 5-(3-Methoxyphenyl)isoxazole