Learn More
5-(2-Fluorophenyl)-1H-pyrrole-3-carbaldehyde, TRC
Product Code. 30133513
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133513 1 g 1g
30009936 250 mg 250mg
30005338 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133513 Supplier Toronto Research Chemicals Supplier No. TRCF4896551G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 881674-56-2
Molecular Formula C11H8FNO
Molecular Weight (g/mol) 189.19
Synonym Vonoprazan Impurity 85
SMILES Fc1ccccc1c2cc(C=O)c[nH]2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C11H8FNO/c12-10-4-2-1-3-9(10)11-5-8(7-14)6-13-11/h1-7,13H
Formula Weight 189.059
Chemical Name or Material 5-(2-Fluorophenyl)-1H-pyrrole-3-carbaldehyde