Learn More
5-(2-Chloroacetyl)indolin-2-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 65435-04-3 |
| Molecular Formula | C10H8ClNO2 |
| MDL Number | MFCD04115734 |
| Quantity | 5 g |
| SMILES | O=C1NC2=C(C=C(C(CCl)=O)C=C2)C1 |
| Molecular Weight (g/mol) | 209.63 |
| Percent Purity | 98% |
| Chemical Name or Material | 5-(2-Chloroacetyl)indolin-2-one |