Learn More
(4S)-4-(Phenylthio)-L-proline Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (4S)-4-(Phenylthio)-L-proline Hydrochloride |
| CAS | 105107-84-4 |
| Molecular Formula | C11H14ClNO2S |
| Quantity | 25 mg |
| Synonym | cis-4-(Phenylthio)-L-proline Hydrochloride |
| InChI Formula | InChI=1S/C11H13NO2S.ClH/c13-11(14)10-6-9(7-12-10)15-8-4-2-1-3-5-8;/h1-5,9-10,12H,6-7H2,(H,13,14);1H/t9-,10-;/m0./s1 |
| SMILES | Cl.OC(=O)[C@@H]1C[C@@H](CN1)Sc2ccccc2 |
| IUPAC Name | (2S,4S)-4-phenylsulfanylpyrrolidine-2-carboxylic acid;hydrochloride |
| Molecular Weight (g/mol) | 259.75 |
| Formula Weight | 259.0434 |