Learn More
(4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester, TRC
Product Code. 30366298
Change view
Click to view available options
Quantity:
500 mg
2.5 g
10 g
Unit Size:
10g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366298 500 mg 500mg
30354917 10 g 10g
30361192 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30366298 Supplier Toronto Research Chemicals Supplier No. TRCC982020500MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 125971-94-0
Molecular Formula C14 H23 N O4
Molecular Weight (g/mol) 269.34
IUPAC Name tert-butyl 2-[(4R,6R)-6-(cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
SMILES CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC#N)OC(C)(C)O1
Specifications
Quantity 500 mg
Synonym 1,3-Dioxane-4-acetic acid, 6-(cyanomethyl)-2,2-dimethyl-, 1,1-dimethylethyl ester, (4R,6R)-, (4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester, : 1,3-Dioxane-4-acetic acid, 6-(cyanomethyl)-2,2-dimethyl-, 1,1-dimethylethyl ester, (4R-cis)-, ((4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-[1,3]dioxan-4-yl)acetic acid tert-butyl ester, (6R-Cyanomethyl-2,2-dimethyl-[1,3]dioxan-4R-yl)acetic acid tert-butyl ester
InChI Formula InChI=1S/C14H23NO4/c1-13(2,3)19-12(16)9-11-8-10(6-7-15)17-14(4,5)18-11/h10-11H,6,8-9H2,1-5H3/t10-,11-/m1/s1
Formula Weight 269.1627
Chemical Name or Material 1,1-Dimethylethyl 2-[(4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate