missing translation for 'onlineSavingsMsg'
Learn More

(4E)-4-Octenedioic Acid-1,8-13C2, TRC

Product Code. 30426331
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426331 1 mg -
30419163 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426331 Supplier Toronto Research Chemicals Supplier No. TRCO2391021MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185234-84-7
Molecular Formula C613C2H12O4
Molecular Weight (g/mol) 174.16
Synonym (E)-4-Octenedioic Acid-1,8-13C2
IUPAC Name (E)-(1,8-^{13}C_{2})oct-4-enedioic acid
SMILES O[13C](=O)CC\C=C\CC[13C](=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C8H12O4/c9-7(10)5-3-1-2-4-6-8(11)12/h1-2H,3-6H2,(H,9,10)(H,11,12)/b2-1+/i7+1,8+1
Formula Weight 174.0803
Chemical Name or Material (4E)-4-Octenedioic Acid-1,8-13C2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.