Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
Model:
1001
1002
1005
1010
1725
1750
1801 RN
1802
1805
1810 RN
1825
701
702
705
710
725
750
75RN
missing translation for 'unitSize'
Each
Description
- PTFE coating from point for 0.75 in.
- For dispensing both liquids and gases
- Precision-machined polymer plunger tip that creates leak-free seal
Available as replacement needle for installing into standard removable needle (RN) syringe; Needles sold separately
Specifications
Specifications
| Needle Gauge | 22 G |
| Product Type | TLC |
| Model | 1750 |
| Volume (Metric) | 500 μL |