Learn More
Linker Industrietechnik™ 400mm Dia. Pure Aluminum Kressner Sieves
Product Code. 11784389
Click to view available options
Diameter (Metric) Outer:
400 mm
Unit Size:
Each
Product Code. 11784389
Brand: Linker Industrietechnik™ 561.407.000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Quantity 1Each
Type Kressner Sieve
Material Aluminum
Includes Lid, 2 sieve rings and 3 adjustable excess pressure catches and collection pan
For Use With (Application) For accommodating exchangeable sieve inserts
Diameter (Metric) Outer 400 mm