Learn More
Linker Industrietechnik™ 400 Dia. x 60 mm Height Stainless Steel Test Sieves
Product Code. 11747686
Click to view available options
Pore Size:
63 μm
1 mm
100 μm
1.4 mm
1.6 mm
2 mm
200 μm
2.5 mm
3.15 mm
315 μm
4 mm
400 μm
425 μm
4.75 mm
5 mm
500 μm
5.6 mm
600 μm
8 mm
850 μm
900 μm
Unit Size:
Each
Product Code. 11747686
Brand: Linker Industrietechnik™ 554.060.222070
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Quantity 1Each
Pore Size 4 mm
Type Test Sieve
Certifications/Compliance Compliance 2.1 EN 10 204 and ISO 3310-1
Material Stainless Steel
Height (Metric) 60mm
Diameter (Metric) Outer 400 mm
Mesh Material Stainless Steel