missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB 40 kDa TRITC-polysucrose Red Fluorophore
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Description
- Product Overview: TRITC-polysucrose is a high molecular weight, water-soluble fluorescent tracer based on polysucrose labeled with tetramethylrhodamine isothiocyanate (TRITC). Offering bright red-orange fluorescence, it is ideal for permeability studies, tracer diffusion experiments, and modeling molecular movement through biological and engineered barriers. TRITC-polysucrose combines excellent biocompatibility with strong, stable fluorescence.
- Features:
- Red-Orange Fluorescence: Excitation maximum at approximately 555 nm and emission maximum at approximately 580 nm, detectable with standard TRITC and rhodamine filter sets.
- High Molecular Weight: Available in a range of sizes, including options such as 20 kDa, 40 kDa, 70 kDa, 100 kDa, and 400 kDa.
- Water-Soluble and Biocompatible: Fully dissolves in aqueous systems without aggregation, suitable for in vitro and in vivo applications.
- Stable TRITC Conjugation: Provides bright, durable fluorescence for live imaging and fixed-sample studies.
- Controlled Labeling: Manufactured under strict quality control to ensure reproducible fluorescence intensity and diffusion properties.
- Benefits:
- Reliable Tracer for Permeability Studies: Tracks the transport of macromolecules across biological and synthetic barriers.
- Ideal for Diffusion and Flow Research: Suitable for use in tissue models, microfluidics, and perfusion systems.
- Supports Live-Cell and Fixed-Tissue Experiments: Bright signal enables dynamic monitoring and endpoint analysis.
- Minimal Non-Specific Binding: Neutral backbone reduces background signal, improving data quality.
- Applications:
- Permeability assays in epithelial, endothelial, and tissue-engineered barriers.
- Tracer diffusion studies in microfluidics and hydrogel systems.
- Vascular leakage and tissue penetration modeling.
- Live and fixed imaging for macromolecule tracking.
- Calibration of flow and perfusion systems.
- Technical Specifications:
- Fluorescence: Excitation maximum approximately 555 nm, emission maximum approximately 580 nm.
- Molecular Weights: Available in a range from 20 kDa up to 400 kDa options.
- Appearance: Light-sensitive, reddish-orange dry powder, fully soluble in water.
- Storage and Stability:
- Store dry at room temperature, protected from moisture and light.
- Stable under recommended conditions for extended periods without significant loss of fluorescence.
- Packaging and Availability:
- Supplied as dry powder for easy handling and long-term storage.
- Available in research-grade packaging sizes to meet varying experimental needs.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | Fluorescent Labeling Reagent |
| Molecular Weight (g/mol) | 40 kDa |
| Label Type | Other Labels or Dyes |
| Color | Red |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction