missing translation for 'onlineSavingsMsg'
Learn More

TdB Labs AB 40 kDa DEAE-dextran for Transfection in Gene Therapy Studies

Product Code. p-200113334
Encompass_Preferred
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
This item is not returnable. View return policy

Product Code. 30216513

Brand: TdB Labs AB DD40_10g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DEAE-dextran 40 kDa is a cationic polysaccharide widely used in transfection, viral enhancement assays, and nucleic acid delivery. Its balance of size and charge makes it a versatile tool in cell culture and biochemical interaction studies.

  • Product Overview: DEAE-dextran is a cationic, water-soluble polysaccharide modified with diethylaminoethyl groups. Known for its strong positive charge and high biocompatibility, DEAE-dextran is ideal for researchers and bioprocess engineers working with protein purification, nucleic acid complexation, or surface functionalization. Available across a wide molecular weight range to match different project needs.
  • Key Features:
    • Strong Cationic Properties: DEAE-modification provides consistent positive charge for robust binding to negatively charged biomolecules like proteins, DNA, and RNA.
    • Flexible Molecular Weight Range: Offered from 4 kDa up to 2000 kDa, giving options for fine-tuning binding strength, diffusion, and separation in your application.
    • High Solubility: Fully water-soluble powder allows rapid and easy preparation for chromatography, formulation, or complexation protocols.
    • Batch-to-Batch Consistency: Tight control over molecular weight and functionalization ensures reliable results across experiments and production runs.
    • Stable Storage: Dry powder format provides long shelf life without special storage conditions, helping to minimize lab downtime.
  • Why Choose DEAE-dextran:
    • For Protein Purification: DEAE-dextran is a proven choice for anion-exchange chromatography workflows, helping isolate or fractionate proteins under mild, biocompatible conditions.
    • For Complexation and Binding Studies: Ideal for creating complexes with nucleic acids or proteins to study binding kinetics, surface adsorption, or biomolecular interactions.
    • For Transfection Research: Historically used in mammalian cell DNA transfection experiments, DEAE-dextran still finds niche use where simple, non-viral gene delivery is studied.
    • For Surface Modification: Researchers functionalizing surfaces (e.g., biosensors, biomaterials) benefit from DEAE-dextran's ability to introduce stable, positive surface charges.
    • For Vaccine Adjuvant Studies: Studied as a component in veterinary vaccine formulations to enhance immune system targeting by improving antigen presentation.
  • Typical Applications:
    • Ion-exchange chromatography for proteins and nucleic acids
    • Formation of electrostatic complexes for analytical and purification purposes
    • Historical and research-based DNA transfection in mammalian cells
    • Surface coating and modification in bioengineering projects
    • Supportive studies in vaccine and adjuvant formulation development
  • Technical Details:
    • Molecular Weights: 4 kDa, 20 kDa, 40 kDa, 70 kDa, 150 kDa, 500 kDa, and 2000 kDa available.
    • Charge Density: Approximately 0.1 to 0.2 DEAE groups per glucose unit (depending on the grade).
    • Appearance: White to off-white dry powder, highly soluble in water.
  • Storage and Handling:
    • Store dry at room temperature in a sealed container to maintain stability.
    • Protect from moisture to preserve full activity over long-term storage.
  • Availability:
    • Supplied as research-grade powder, suitable for laboratory use and pilot-scale process development.
    • Available in small packs for R and D or bulk volumes for production environments.

Specifications

Cell Type HEK293
Format Vial
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Species Human
Product Type Transfection Reagent
Quantity 10 g
Sterility Non-sterile
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.