Learn More
Thermo Scientific™ 4-Way Flipper™ Racks
GREENER_CHOICE
Product Code. 11520125 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Color:
Fluorescent Green
Fluorescent Orange
Fluorescent Pink
Unit Size:
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11520125 Fluorescent Orange Pack of 20
11530125 Fluorescent Pink Pack of 20
10321031 Fluorescent Green Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11520125 Supplier Thermo Scientific™ Supplier No. 8860
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™4-Way Flipper™ Racks.
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™ 4-Way Flipper™ Racks.
TRUSTED_SUSTAINABILITY
Specifications
Description 4-Way Flipper Rack
Color Fluorescent Orange
Holds 0.5 mL, 15 mL, Tubes (50 mL)
Positions 32 x 0.5 mL, 32 x 1.5 mL, 12 x 15 mL or 4 x 50 mL tubes, each on a different side
Shape Rectangular
Product Type 4-way Flipper Rack