Learn More
4-(Trifluoromethylthio)benzoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(Trifluoromethylthio)benzoyl Chloride |
| CAS | 330-14-3 |
| Molecular Formula | C8H4ClF3OS |
| Quantity | 5 g |
| Synonym | 4-Trifluoromethylsulfanylbenzoyl Chloride, p-[(Trifluoromethyl)thio]benzoyl Chloride, Benzoyl chloride','p-[(trifluoromethyl)thio], 4-[(Trifluoromethyl)thio]benzoyl chloride |
| InChI Formula | InChI=1S/C8H4ClF3OS/c9-7(13)5-1-3-6(4-2-5)14-8(10,11)12/h1-4H |
| SMILES | ClC(C1=CC=C(SC(F)(F)F)C=C1)=O |
| IUPAC Name | 4-((trifluoromethyl)thio)benzoyl chloride |
| Molecular Weight (g/mol) | 240.63 |
| Formula Weight | 239.96 |