Learn More
4-(Trifluoromethyl)phenylacetic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 32857-62-8 |
| Molecular Formula | C9H7F3O2 |
| MDL Number | MFCD00004352 |
| Quantity | 100 g |
| SMILES | C1=C(C=CC(=C1)CC(O)=O)C(F)(F)F |
| Molecular Weight (g/mol) | 204.15 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-(Trifluoromethyl)phenylacetic acid |