Learn More
(4-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 2923-56-0 |
| Molecular Formula | C7H8ClF3N2 |
| MDL Number | MFCD00204233 |
| Quantity | 5 g |
| SMILES | Cl.FC(C1=CC=C(C=C1)NN)(F)F |
| Molecular Weight (g/mol) | 212.60 |
| Percent Purity | 97% |
| Chemical Name or Material | (4-(Trifluoromethyl)phenyl)hydrazine hydrochloride |