missing translation for 'onlineSavingsMsg'
Learn More

[4-(Trifluoromethoxy)phenyl]carbamic Acid Methyl Ester, TRC

Product Code. 30456981
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456981 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456981 Supplier Toronto Research Chemicals Supplier No. TRCT79075025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [4-(Trifluoromethoxy)phenyl]carbamic acid methyl ester
CAS 177905-10-1
Molecular Formula C9 H8 F3 N O3
Quantity 25 mg
Synonym Carbamic acid','N-[4-(trifluoromethoxy)phenyl]-','methyl ester, Carbamic acid','[4-(trifluoromethoxy)phenyl]-','methyl ester (9CI), IN-KB 687
InChI Formula InChI=1S/C9H8F3NO3/c1-15-8(14)13-6-2-4-7(5-3-6)16-9(10,11)12/h2-5H,1H3,(H,13,14)
SMILES COC(=O)Nc1ccc(OC(F)(F)F)cc1
IUPAC Name methyl N-[4-(trifluoromethoxy)phenyl]carbamate
Molecular Weight (g/mol) 235.16
Formula Weight 235.0456
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.