missing translation for 'onlineSavingsMsg'
Learn More

4-Thiocyanato-O-cresol, TRC

Product Code. 30450460
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450460 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450460 Supplier Toronto Research Chemicals Supplier No. TRCT3444002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Thiocyanato-O-cresol
CAS 3774-53-6
Molecular Formula C8H7NOS
Quantity 2.5 g
Synonym Thiocyanic Acid 4-Hydroxy-3-methylphenyl Ester, 2-Methyl-4-(thiocyano)phenol, 4-Hydroxy-3-methylphenyl Thiocyanate, 4-Hydroxy-m-tolyl Thiocyanate, NSC 31040, NSC 4470, NSC 7392
InChI Formula InChI=1S/C8H7NOS/c1-6-4-7(11-5-9)2-3-8(6)10/h2-4,10H,1H3
SMILES Cc1cc(SC#N)ccc1O
IUPAC Name (4-hydroxy-3-methylphenyl) thiocyanate
Molecular Weight (g/mol) 165.21
Formula Weight 165.0248
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.