Learn More
4-(tert-Butyl)benzene-1,2-diamine, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 68176-57-8 |
| Molecular Formula | C10H16N2 |
| MDL Number | MFCD00052695 |
| Quantity | 5 g |
| SMILES | NC1=CC=C(C(C)(C)C)C=C1N |
| Molecular Weight (g/mol) | 164.25 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(tert-Butyl)benzene-1,2-diamine |