Learn More
4-Pyridinealdoxime, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Pyridinealdoxime |
| CAS | 696-54-8 |
| Molecular Formula | C6 H6 N2 O |
| Quantity | 250 mg |
| Synonym | 4-Pyridinecarboxaldehyde','oxime, 4-Pyridinecarboxaldehyde oxime, Isonicotinaldehyde','oxime (6CI,8CI), 4-(Hydroxyiminomethyl)pyridine, 4-Pyridinaldoxime, 4-Pyridinealdoxime, NSC 63847, Pyridine-4-carboxaldoxime |
| InChI Formula | InChI=1S/C6H6N2O/c9-8-5-6-1-3-7-4-2-6/h1-5,9H/b8-5+ |
| SMILES | O\N=C\c1ccncc1 |
| IUPAC Name | (NE)-N-(pyridin-4-ylmethylidene)hydroxylamine |
| Molecular Weight (g/mol) | 122.12 |
| Formula Weight | 122.048 |