Learn More
4-Pyridin-4-yl-benzoic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4385-76-6 |
| Molecular Formula | C12H9NO2 |
| MDL Number | MFCD04114574 |
| Quantity | 25 g |
| SMILES | N1=CC=C(C=C1)C2=CC=C(C(=O)O)C=C2 |
| Molecular Weight (g/mol) | 199.21 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Pyridin-4-yl-benzoic acid |