missing translation for 'onlineSavingsMsg'
Learn More

4-(pyrid-2-yloxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10092364
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10092364 5 g 5g
10652325 250 mg 250mg
10662325 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092364 Supplier Thermo Scientific Maybridge Supplier No. CC48904DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 194017-69-1
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
InChI Key DPRZACGKYIDYCK-UHFFFAOYSA-N
Synonym 4-pyridin-2-yloxy benzaldehyde, 4-pyrid-2-yloxy benzaldehyde, 4-2-pyridyloxy benzaldehyde, benzaldehyde,4-2-pyridinyloxy, 4-2-pyridinyloxy benzaldehyde, 4-2-pyridinyloxy-benzaldehyde, 4-pyridin-2-yloxy-benzaldehyde, 4-2-pyridyloxy phenyl formaldehyde
PubChem CID 3755292
IUPAC Name 4-pyridin-2-yloxybenzaldehyde
SMILES C1=CC=NC(=C1)OC2=CC=C(C=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 4-(Pyrid-2-yloxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.