missing translation for 'onlineSavingsMsg'
Learn More

4-Propionamidophenol (Acetaminophen Impurity B), TRC

Product Code. 17952225
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952225 25 mg 25mg
30134835 100 mg 100mg
17911175 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952225 Supplier Toronto Research Chemicals Supplier No. TRCP78350025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1693-37-4
Molecular Formula C9 H11 N O2
Molecular Weight (g/mol) 165.19
Synonym Propanamide, N-(4-hydroxyphenyl)-, Propionanilide, 4'-hydroxy- (7CI,8CI), N-(4-Hydroxyphenyl)propanamide, 4-(Propionylamino)phenol, 4-Propionamidophenol, 4'-Hydroxypropionanilid, 4'-Hydroxypropionanilide, N-(4-Hydroxyphenyl)propionamide, N-(p-Hydroxyphenyl)propionamide, Parapropamol, Parapropanol, Solvodol, p-Hydroxypropionanilide, p-Propionamidophenol, N-Propionyl-4-aminophenol, Paracetamol Imp. B (EP)
IUPAC Name N-(4-hydroxyphenyl)propanamide
SMILES CCC(=O)Nc1ccc(O)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H11NO2/c1-2-9(12)10-7-3-5-8(11)6-4-7/h3-6,11H,2H2,1H3,(H,10,12)
Formula Weight 165.079
Percent Purity >95%
Chemical Name or Material 4-Propionamidophenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.